Learn More
Cayman Chemical ssK36 (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502915687
Encompass_Preferred
A peptide super-substrate for SETD2; methylated 70-fold more and 290-fold faster than H3K36 (29-43) in cell-free assays
Catalog No. 50-291-5687 Supplier Cayman Chemical Supplier No. 3751410 MG
A peptide super-substrate for SETD2; methylated 70-fold more and 290-fold faster than H3K36 (29-43) in cell-free assays