Learn More
Medchemexpress LLC HY-107414 5mg Medchemexpress, SKA-121 CAS:1820708-73-3 Purity:>98%
SDP
Catalog No. 501968405
Encompass_Preferred
Medchemexpress, HY-107414 5mg SKA-121 CAS:1820708-73-3 SKA-121 is a selective KCa3.1 activator. SKA-121 exhibits EC50s of 109 nM and 4.4 μM for KCa3.1 and KCa2.3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8405 Supplier Medchemexpress LLC Supplier No. HY1074145MG
Medchemexpress, HY-107414 5mg SKA-121 CAS:1820708-73-3 SKA-121 is a selective KCa3.1 activator. SKA-121 exhibits EC50s of 109 nM and 4.4 μM for KCa3.1 and KCa2.3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.