Learn More
Medchemexpress LLC SKA-31 | 40172-65-4 | 99.7% | 200.27 | 1 ML
SDP
Catalog No. 5000439879
Encompass_Preferred
SKA-31 is a potent potassium channel activator that demonstrates significant activity against KCa3.1, KCa2.2, KCa2.1, and KCa2.3 channels. It potentiates endothelium-derived hyperpolarizing factor response and has been shown to lower blood pressure in studies. This compound is intended for research use only.
  • Activates KCa2/3 channels potently and selectively
  • Reduces cell viability in HCT-116 and HCT-8 cells
  • Triggers apoptosis in HCT-116 cells
  • Increases cells in G0/G1 phase
  • Potentiates endothelium-derived hyperpolarizing factor response
  • Lowers blood pressure in mice
Catalog No. 50-004-39879 Supplier Medchemexpress LLC Supplier No. HY11165510MM/1ML
SKA-31 is a potent potassium channel activator that demonstrates significant activity against KCa3.1, KCa2.2, KCa2.1, and KCa2.3 channels. It potentiates endothelium-derived hyperpolarizing factor response and has been shown to lower blood pressure in studies. This compound is intended for research use only.
  • Activates KCa2/3 channels potently and selectively
  • Reduces cell viability in HCT-116 and HCT-8 cells
  • Triggers apoptosis in HCT-116 cells
  • Increases cells in G0/G1 phase
  • Potentiates endothelium-derived hyperpolarizing factor response
  • Lowers blood pressure in mice