Learn More
Scott Safety™ SKA-PAK™ Automatic Transfer Supplied Air Respirator
Catalog No. 19121861
Change view
Click to view available options
Duration:
15 min.
Includes:
Dupont™ Kevlar™ aramid fiber harness, 4500psi carbon 15 min. cylinder, no facepiece, E-Z FLO Vibralert QD regulator, Schrader airline fitting
Catalog No. Duration Includes
19-121-861 15 min. Dupont™ Kevlar™ aramid fiber harness, 4500psi carbon 15 min. cylinder, no facepiece, E-Z FLO Vibralert QD regulator, Schrader airline fitting
1 options
Catalog No. 19-121-861 Supplier Scott Safety™ Supplier No. 7012623470
Please contact your local Fisher Scientific sales representative for pricing
Pressure demand supplied air respirator equipped with an automatic, hands-free air transfer capability.
  • Combination Type C positive pressure respirator
  • Automatic, hands-free transfer from external air source to cylinder in the event of primary air source interruption
  • Personal, tactile Vibralert alarm to alert users of transfer to cylinder air and enable prompt escape
  • Onboard cylinder gauge and 90% air remaining lighted alarm for added safety and awareness
  • E-Z Klip cylinder retention system for quick and easy cylinder changes
Specifications
Certifications/Compliance NIOSH
Product Type Automatic Transfer Supplied Air Respirator
Duration 15 min.
For Use With (Application) Confined space, hazardous materials clean-up, chemical, petrochemical, oil and gas, and public utilities.
Includes Dupont™ Kevlar™ aramid fiber harness, 4500psi carbon 15 min. cylinder, no facepiece, E-Z FLO Vibralert QD regulator, Schrader airline fitting
Model SAR424060442001