Learn More
Aviva Systems Biology Corp Ska1 Antibody - middle region (ARP55460_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55460_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ska1. It was validated on Western Blot. . Alternate names: 2810433K01Rik, AV117428ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-040