Learn More
Aviva Systems Biology Corp SKA3 Antibody - middle region (ARP55463_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55463_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C13orf3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SKA3. Alternate names: MGC4832, RAMA1, C13orf3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-53870