Learn More
Aviva Systems Biology Corp SKAP1 Antibody - N-terminal region (ARP46047_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46047_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SKAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SKAP1. Alternate names: SCAP1, SKAP55ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59196