Learn More
Medchemexpress LLC Skatole (3-methylindole) | 83-34-1 | 131.17 | 50 G
SDP
Catalog No. 5000460249
Encompass_Preferred
Skatole (3-methylindole) is a heterocyclic compound that occurs naturally in the feces of vertebrates and in certain plants. It can be produced by intestinal bacteria, where it induces apoptosis of intestinal epithelial cells by activating aryl hydrocarbon receptors (AhR) and p38. Skatole has applications in specific products within the perfume industry and as a flavor additive in ice cream.
  • Naturally found in the feces of vertebrates and certain plants.
  • Produced by intestinal bacteria.
  • Induces apoptosis of intestinal epithelial cells through activating aryl hydrocarbon receptors (AhR) and p38.
  • Used in specific products of the perfume industry.
  • Used as a flavor additive in ice cream.
Catalog No. 50-004-60249 Supplier Medchemexpress LLC Supplier No. HYW00735550G
Add to Cart
Add to Cart
Skatole (3-methylindole) is a heterocyclic compound that occurs naturally in the feces of vertebrates and in certain plants. It can be produced by intestinal bacteria, where it induces apoptosis of intestinal epithelial cells by activating aryl hydrocarbon receptors (AhR) and p38. Skatole has applications in specific products within the perfume industry and as a flavor additive in ice cream.
  • Naturally found in the feces of vertebrates and certain plants.
  • Produced by intestinal bacteria.
  • Induces apoptosis of intestinal epithelial cells through activating aryl hydrocarbon receptors (AhR) and p38.
  • Used in specific products of the perfume industry.
  • Used as a flavor additive in ice cream.