Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Skatole (3-Methylindole) | 83-34-1 | MFCD00005627 | 131.17 | 100 G
SDP

Catalog No. 5000460338
Encompass_Preferred

Skatole (3-Methylindole) is a heterocyclic compound naturally occurring in vertebrate feces and some plants. Produced by intestinal bacteria, it can induce apoptosis in intestinal epithelial cells by activating aryl hydrocarbon receptors (AhR) and p38. This compound finds utility in the perfume industry and as a flavor additive in ice cream.

  • Purity of 99.92%
  • Molecular weight of 131.17
  • Molecular formula: C9H9N
  • Solid appearance
  • White to light brown color
  • Derived from endogenous metabolites, human gut microbiota metabolites, and microorganisms
  • Used in the perfume industry
  • Functions as a flavor additive in ice cream
  • Induces apoptosis in intestinal epithelial cells by activating aryl hydrocarbon receptors (AhR) and p38
  • Reverses suppression of Irisin on Ang II-evoked proliferation, migration, and phenotypic transformation in VSMCs

Catalog No. 50-004-60338 Supplier Medchemexpress LLC Supplier No. HYW007355100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Skatole (3-Methylindole) is a heterocyclic compound naturally occurring in vertebrate feces and some plants. Produced by intestinal bacteria, it can induce apoptosis in intestinal epithelial cells by activating aryl hydrocarbon receptors (AhR) and p38. This compound finds utility in the perfume industry and as a flavor additive in ice cream.

  • Purity of 99.92%
  • Molecular weight of 131.17
  • Molecular formula: C9H9N
  • Solid appearance
  • White to light brown color
  • Derived from endogenous metabolites, human gut microbiota metabolites, and microorganisms
  • Used in the perfume industry
  • Functions as a flavor additive in ice cream
  • Induces apoptosis in intestinal epithelial cells by activating aryl hydrocarbon receptors (AhR) and p38
  • Reverses suppression of Irisin on Ang II-evoked proliferation, migration, and phenotypic transformation in VSMCs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.