Learn More
Medchemexpress LLC Skatole-d3 | 111399-60-1 | 134.19 | 10 MG
SDP
Catalog No. 5000460322
Encompass_Preferred
Skatole-d3 is a deuterium labeled form of Skatole. Skatole is produced by intestinal bacteria and regulates intestinal epithelial cellular functions by activating aryl hydrocarbon receptors and p38. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. It is identified for use as a laboratory chemical and in the manufacture of substances.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Laboratory chemical
  • Used in the manufacture of substances
Catalog No. 50-004-60322 Supplier Medchemexpress LLC Supplier No. HYW007355S10MG
Skatole-d3 is a deuterium labeled form of Skatole. Skatole is produced by intestinal bacteria and regulates intestinal epithelial cellular functions by activating aryl hydrocarbon receptors and p38. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. It is identified for use as a laboratory chemical and in the manufacture of substances.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Laboratory chemical
  • Used in the manufacture of substances