Learn More
Medchemexpress LLC Skatole-d8 (3-Methylindole-d8) | 697807-03-7 | 139.22 | 5 MG
SDP
Catalog No. 5000460303
Encompass_Preferred
Skatole-d8 is the deuterium labeled Skatole. Produced by intestinal bacteria, Skatole regulates intestinal epithelial cellular functions through activating aryl hydrocarbon receptors and p38. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-60303 Supplier Medchemexpress LLC Supplier No. HYW007355S15MG
Skatole-d8 is the deuterium labeled Skatole. Produced by intestinal bacteria, Skatole regulates intestinal epithelial cellular functions through activating aryl hydrocarbon receptors and p38. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS