Learn More
Medchemexpress LLC Skatole (standard) | 83-34-1 | 131.17 | 250 MG
SDP
Catalog No. 5000449824
Encompass_Preferred
Skatole (Standard) is the analytical standard of Skatole, used for research and analytical applications. It is a heterocyclic compound, 3-methylindole, produced by intestinal bacteria, and has applications in the perfume industry and as a flavor additive in ice cream. It can also induce apoptosis of intestinal epithelial cells by activating aryl hydrocarbon receptors (AhR) and p38.
  • Analytical standard for Skatole
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-49824 Supplier Medchemexpress LLC Supplier No. 250MGHYW007355R
Skatole (Standard) is the analytical standard of Skatole, used for research and analytical applications. It is a heterocyclic compound, 3-methylindole, produced by intestinal bacteria, and has applications in the perfume industry and as a flavor additive in ice cream. It can also induce apoptosis of intestinal epithelial cells by activating aryl hydrocarbon receptors (AhR) and p38.
  • Analytical standard for Skatole
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS