Learn More
MSC SKB Corporation Clamshell Case, Cubed Foam, 18.5 x 13 x 5.5 in, Polyethylene
Catalog No. 5000528545
Encompass
This SKB Corporation Clamshell Case is a black polyethylene protective transport case featuring cubed foam. It measures 18.5 inches in length, 13 inches in width, and 5.5 inches in depth. Designed for durability, it includes a padlockable locking mechanism and a cushion grip, folding handle.
  • Patented trigger release latches
  • Resistant to UV, solvents, corrosion, fungus & impact damage (MIL-STD 810H)
  • Waterproof & dust tight design (MIL-STD 810H)
  • Molded-in hinge for added protection
  • Snap-down rubber over-molded cushion grip handle
  • Automatic ambient pressure equalization valve
  • Padlock loop
  • Cubed foam
Catalog No. 50-005-28545 Supplier MSC Supplier No. 51540946
This SKB Corporation Clamshell Case is a black polyethylene protective transport case featuring cubed foam. It measures 18.5 inches in length, 13 inches in width, and 5.5 inches in depth. Designed for durability, it includes a padlockable locking mechanism and a cushion grip, folding handle.
  • Patented trigger release latches
  • Resistant to UV, solvents, corrosion, fungus & impact damage (MIL-STD 810H)
  • Waterproof & dust tight design (MIL-STD 810H)
  • Molded-in hinge for added protection
  • Snap-down rubber over-molded cushion grip handle
  • Automatic ambient pressure equalization valve
  • Padlock loop
  • Cubed foam