Learn More
MSC SKB Corporation iSeries 3424-12 Case, Watertight, Cubed Foam, 34.5 x 24.5 x 12.75 in
Catalog No. 5000533938
Encompass
SKB iSeries Injection Molded Mil-Standard Waterproof Cases are constructed from ultra high-strength polypropylene copolymer resin. These cases feature a gasket-sealed, water and dust tight, submersible design (MIL-STD 810H) that resists corrosion and impact damage.
  • Oversized heavy-duty handles and wide-set double wheels
  • UV, solvent, corrosion, and fungus resistant (MIL-STD 810H)
  • Complete gasket seal for watertight protection (MIL-STD 810H)
  • Automatic ambient pressure equalization valve (MIL-STD 810H)
  • Molded-in hinges
  • Patented "trigger release" TSA locking latch system
  • TSA recognized and accepted locks can be retrofitted
  • Stainless steel locking loops for customer supplied lock
  • Cubed foam interior for customization
Catalog No. 50-005-33938 Supplier MSC Supplier No. 19913896
SKB iSeries Injection Molded Mil-Standard Waterproof Cases are constructed from ultra high-strength polypropylene copolymer resin. These cases feature a gasket-sealed, water and dust tight, submersible design (MIL-STD 810H) that resists corrosion and impact damage.
  • Oversized heavy-duty handles and wide-set double wheels
  • UV, solvent, corrosion, and fungus resistant (MIL-STD 810H)
  • Complete gasket seal for watertight protection (MIL-STD 810H)
  • Automatic ambient pressure equalization valve (MIL-STD 810H)
  • Molded-in hinges
  • Patented "trigger release" TSA locking latch system
  • TSA recognized and accepted locks can be retrofitted
  • Stainless steel locking loops for customer supplied lock
  • Cubed foam interior for customization