Learn More
Lonza Walkersville SKBM- 2 Skeletal Muscle Cell Growth Basal Medium-2
Catalog No. NC9992029
Encompass
SkBM -2 is a Skeletal Muscle Myoblast basal medium sold separately. This can be used with 1 kit of CC-3244 (skeletal muscle myoblast growth medium SingleQuots Supplements and Growth Factors). The full SkGM -2 BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3245. We offers two varieties of Skeletal Muscle Growth Medium - SkGM and SkGM-2.SkGM-2 Skeletal Muscle Cell Growth Medium-2 (catalog number CC-3245) is an enhancement of SkGM Medium. Fetuin has been removed and there has been further optimization of the basal media.SkGM Medium is optimized for use with human skeletal muscle cells. SkGM -2 is specially designed for use with normal and diabetic skeletal muscle myoblasts.Content:1 x Bottle Basal Medium 500 mL
Catalog No. NC9992029 Supplier Lonza Walkersville Supplier No. CC3246
SkBM -2 is a Skeletal Muscle Myoblast basal medium sold separately. This can be used with 1 kit of CC-3244 (skeletal muscle myoblast growth medium SingleQuots Supplements and Growth Factors). The full SkGM -2 BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3245. We offers two varieties of Skeletal Muscle Growth Medium - SkGM and SkGM-2.SkGM-2 Skeletal Muscle Cell Growth Medium-2 (catalog number CC-3245) is an enhancement of SkGM Medium. Fetuin has been removed and there has been further optimization of the basal media.SkGM Medium is optimized for use with human skeletal muscle cells. SkGM -2 is specially designed for use with normal and diabetic skeletal muscle myoblasts.Content:1 x Bottle Basal Medium 500 mL