Learn More
Amsbio LLC Skeletal Muscle Cell Differentiation Medium| 500ml
Catalog No. 502563587
Encompass
Skeletal Muscle Cell Differentiation Medium 500ml for myoblast into myocytes. Componenets: DMEM (1g/L D-glucose), Horse serum, Bovine Serum Albumin, Fetuin, Penicillin , Streptomycin, Amphotericin B.
Catalog No. 50-256-3587 Supplier Amsbio LLC Supplier No. SKMD
Skeletal Muscle Cell Differentiation Medium 500ml for myoblast into myocytes. Componenets: DMEM (1g/L D-glucose), Horse serum, Bovine Serum Albumin, Fetuin, Penicillin , Streptomycin, Amphotericin B.