Learn More
Amsbio LLC Skeletal Muscle Cell Growth Medium| 500ml
Catalog No. 502563584
Encompass
Skeletal Muscle Cell Growth Medium 500ml for myoblast culture. Components: DMEM (1g/L D-glucose), Fetal Bovine Serum, Bovine Serum Albumin, Fetuin, Human Epidermal Growth Factor (hEGF), Dexamethasone, Human Insulin, Penicillin, Streptomycin, Amphotericin B.
Catalog No. 50-256-3584 Supplier Amsbio LLC Supplier No. SKMM
Skeletal Muscle Cell Growth Medium 500ml for myoblast culture. Components: DMEM (1g/L D-glucose), Fetal Bovine Serum, Bovine Serum Albumin, Fetuin, Human Epidermal Growth Factor (hEGF), Dexamethasone, Human Insulin, Penicillin, Streptomycin, Amphotericin B.