Learn More
Sciencell Research Laboratories Skeletal Muscle Cell Medium, 500 ml, Low-serum (5% V/V FBS), Contains essential amino acids, vitamins, hormones, and growth factors
Catalog No. NC0928077
Encompass
The ScienCell Research Laboratories Inc. Skeletal Muscle Cell Medium is a specialized, low-serum growth medium designed to enhance the proliferation and maintenance of primary human skeletal muscle cells. This medium not only supports human cells but also accommodates cultures from murine, rat, canine, and rabbit species. It arrives as a complete kit, providing a stable environment for cell growth and is tailored to minimize variability through its low serum content. In addition, the formulation incorporates essential components for optimal cell health.
  • Optimized growth for primary human skeletal muscle cells.
  • Compatible with murine, rat, canine, and rabbit skeletal muscle cells.
  • Low-serum formulation (5% V/V fetal bovine serum) enhances experimental control.
  • Available as ready-to-use solutions and customizable kits.
  • Comprehensive nutrient profile with amino acids, vitamins, and growth factors.
Catalog No. NC0928077 Supplier Sciencell Research Laboratories Supplier No. 3501
Add to Cart
Edge
Add to Cart
The ScienCell Research Laboratories Inc. Skeletal Muscle Cell Medium is a specialized, low-serum growth medium designed to enhance the proliferation and maintenance of primary human skeletal muscle cells. This medium not only supports human cells but also accommodates cultures from murine, rat, canine, and rabbit species. It arrives as a complete kit, providing a stable environment for cell growth and is tailored to minimize variability through its low serum content. In addition, the formulation incorporates essential components for optimal cell health.
  • Optimized growth for primary human skeletal muscle cells.
  • Compatible with murine, rat, canine, and rabbit skeletal muscle cells.
  • Low-serum formulation (5% V/V fetal bovine serum) enhances experimental control.
  • Available as ready-to-use solutions and customizable kits.
  • Comprehensive nutrient profile with amino acids, vitamins, and growth factors.