Learn More
ATCC Skeletal Muscle Differentiation Tool
Catalog No. 502384399
Encompass
The Skeletal Muscle Differentiation Tool is a single-component media for the differentiation of Primary Skeletal Muscle Cells. Muscle Cells can be expanded in vitro with complete media before final differentiation into Skeletal Muscle Cells.
Catalog No. 50-238-4399 Supplier ATCC Supplier No. PCS950050
The Skeletal Muscle Differentiation Tool is a single-component media for the differentiation of Primary Skeletal Muscle Cells. Muscle Cells can be expanded in vitro with complete media before final differentiation into Skeletal Muscle Cells.