Learn More
Lifeline Cell Technology Skeletal Muscle Media Kit
SDP
Supplier:  Lifeline Cell Technology LL0069
Encompass_Preferred
StemLife Sk Medium Complete Kit. Culture medium for the growth of skeletal muscle cells. LM-0011 StemLife Basal Medium and LS-1103 StemLife SK LifeFactors Kit
Catalog No. NC1944652