Learn More
Medchemexpress LLC HY-128923 5mg Medchemexpress, SKF-34288 (hydrochloride) CAS:320386-54-7 Purity:>98%
SDP
Catalog No. 501969785
Encompass_Preferred
Medchemexpress, HY-128923 5mg SKF-34288 (HCl) CAS:320386-54-7 SKF-34288 hydrochloride (3-Mercaptopicolinic acid) is a phosphoenolpyruvate carboxykinase (PEPCK) inhibitor.SKF-34288 hydrochloride is a potent hypoglycemic agent via inhibition of glucose synthesis through the specific inhibition of PEPCK in the gluconeogenesis pathway. SKF-34288 hydrochloride inhibits Asn metabolism and results in an increase in amino acids and amides. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9785 Supplier Medchemexpress LLC Supplier No. HY1289235MG
Medchemexpress, HY-128923 5mg SKF-34288 (HCl) CAS:320386-54-7 SKF-34288 hydrochloride (3-Mercaptopicolinic acid) is a phosphoenolpyruvate carboxykinase (PEPCK) inhibitor.SKF-34288 hydrochloride is a potent hypoglycemic agent via inhibition of glucose synthesis through the specific inhibition of PEPCK in the gluconeogenesis pathway. SKF-34288 hydrochloride inhibits Asn metabolism and results in an increase in amino acids and amides. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.