Learn More
Medchemexpress LLC SKF-34288 hydrochloride (3-Mercaptopicolinic acid hydrochloride) | 320386-54-7 | 98.6% | 100 MG
SDP
Catalog No. 5000338361
Encompass_Preferred
SKF-34288 hydrochloride (3-Mercaptopicolinic acid) is an orally active phosphoenolpyruvate carboxykinase (PEPCK) inhibitor with a Ki of 2-9 μM. It acts as a potent hypoglycemic agent by inhibiting glucose synthesis and also inhibits Asn metabolism, leading to an increase in amino acids and amides.
  • Orally active PEPCK inhibitor (Ki: 2-9 μM)
  • Potent hypoglycemic agent that inhibits glucose synthesis
  • Inhibits Asn metabolism
  • Increases amino acids and amides
  • Reduces blood glucose in starved rats
  • Neutralizes Salbutamol-mediated hyperglycaemia in rabbits
  • Reduces pyruvate-induced blood glucose level in rats
  • Inhibits C2C12 cell proliferation
  • Induces myogenic differentiation of C2C12 cells
Catalog No. 50-003-38361 Supplier Medchemexpress LLC Supplier No. HY128923100MG
SKF-34288 hydrochloride (3-Mercaptopicolinic acid) is an orally active phosphoenolpyruvate carboxykinase (PEPCK) inhibitor with a Ki of 2-9 μM. It acts as a potent hypoglycemic agent by inhibiting glucose synthesis and also inhibits Asn metabolism, leading to an increase in amino acids and amides.
  • Orally active PEPCK inhibitor (Ki: 2-9 μM)
  • Potent hypoglycemic agent that inhibits glucose synthesis
  • Inhibits Asn metabolism
  • Increases amino acids and amides
  • Reduces blood glucose in starved rats
  • Neutralizes Salbutamol-mediated hyperglycaemia in rabbits
  • Reduces pyruvate-induced blood glucose level in rats
  • Inhibits C2C12 cell proliferation
  • Induces myogenic differentiation of C2C12 cells