Learn More
Medchemexpress LLC Skf-34288 (hydrochloride) | 320386-54-7 | 98.6% | 50 MG
SDP
Catalog No. 5000340441
Encompass_Preferred
SKF-34288 hydrochloride is an orally active phosphoenolpyruvate carboxykinase (PEPCK) inhibitor with a Ki of 2-9 μM. It functions as a potent hypoglycemic agent by inhibiting glucose synthesis. Additionally, it inhibits Asn metabolism, leading to an increase in amino acids and amides.
  • Inhibits C2C12 cell proliferation
  • Induces myogenic differentiation of C2C12 cells
  • Reduces mRNA expression of Pck2 and genes associated with serine biosynthesis
  • Reduces blood glucose in starved rats
  • Neutralizes Salbutamol-mediated hyperglycaemia in rabbits
  • Reduces Pyruvate-induced blood glucose level in rats
Catalog No. 50-003-40441 Supplier Medchemexpress LLC Supplier No. HY12892350MG
SKF-34288 hydrochloride is an orally active phosphoenolpyruvate carboxykinase (PEPCK) inhibitor with a Ki of 2-9 μM. It functions as a potent hypoglycemic agent by inhibiting glucose synthesis. Additionally, it inhibits Asn metabolism, leading to an increase in amino acids and amides.
  • Inhibits C2C12 cell proliferation
  • Induces myogenic differentiation of C2C12 cells
  • Reduces mRNA expression of Pck2 and genes associated with serine biosynthesis
  • Reduces blood glucose in starved rats
  • Neutralizes Salbutamol-mediated hyperglycaemia in rabbits
  • Reduces Pyruvate-induced blood glucose level in rats