Learn More
TARGETMOL CHEMICALS INC SKF-34288 HYDROCHLORIDE 5MG
SDP
Catalog No. 502581395
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. SKF-34288 hydrochloride (3-Mercaptopicolinic acid hydrochloride) is an inhibitor of phosphoenolpyruvate carboxykinase (PEPCK)and inhibits Asn metabolism and results in an increase in amino acids and amides. purity: 95%
Catalog No. 50-258-1395 Supplier TARGETMOL CHEMICALS INC Supplier No. T129295MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. SKF-34288 hydrochloride (3-Mercaptopicolinic acid hydrochloride) is an inhibitor of phosphoenolpyruvate carboxykinase (PEPCK)and inhibits Asn metabolism and results in an increase in amino acids and amides. purity: 95%