Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SKF 38393 hydrochloride | 62717-42-4 | 99.5% | 291.77 | C16H18ClNO2 | 10MM 1ML
SDP

Catalog No. 5000237681
Encompass_Preferred

SKF 38393 hydrochloride is a selective agonist of D1-like dopamine receptors used in pharmacological research. It is provided as a ready-to-use 10 mM solution in DMSO or as a solid for custom preparation, has molecular formula C16H18ClNO2 and molecular weight 291.77 g/mol, and is reported with an IC50 of 110 nM and high purity for analytical work.

  • Selective D1 receptor agonist with reported IC50 of 110 nM.
  • Supplied as 10 mM solution in DMSO or as solid for custom preparation.
  • High purity typically around 99.5% by HPLC.
  • Molecular weight 291.77 g/mol.
  • Chemical formula C16H18ClNO2.
  • Intended for research use only.

Catalog No. 50-002-37681 Supplier Medchemexpress LLC Supplier No. HY12520A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SKF 38393 hydrochloride is a selective agonist of D1-like dopamine receptors used in pharmacological research. It is provided as a ready-to-use 10 mM solution in DMSO or as a solid for custom preparation, has molecular formula C16H18ClNO2 and molecular weight 291.77 g/mol, and is reported with an IC50 of 110 nM and high purity for analytical work.

  • Selective D1 receptor agonist with reported IC50 of 110 nM.
  • Supplied as 10 mM solution in DMSO or as solid for custom preparation.
  • High purity typically around 99.5% by HPLC.
  • Molecular weight 291.77 g/mol.
  • Chemical formula C16H18ClNO2.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.