Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SKF 38393 hydrochloride | 62717-42-4 | MFCD00069248 | >98.0% | 291.77 g/mol | C16H18ClNO2 | 200MG
SDP

Catalog No. 5000238719
Encompass_Preferred

SKF 38393 hydrochloride is the hydrochloride salt of SKF-38393, a selective dopamine D1 receptor agonist with a reported IC50 of approximately 110 nM. Supplied as a research-grade solid analytical standard, it is used for in vitro pharmacology, receptor binding, and analytical assays. Not for human or clinical use.

  • Selective D1 receptor agonist, reported IC50 ≈ 110 nM.
  • Hydrochloride salt form improves solubility for biological assays.
  • Molecular weight 291.77 g/mol.
  • Chemical formula C16H18ClNO2.
  • Provided as a solid analytical standard suitable for in vitro studies.
  • Typical purities reported ≥98% by HPLC from third-party suppliers.
  • Not for human or clinical use; for research only.

Catalog No. 50-002-38719 Supplier Medchemexpress LLC Supplier No. HY12520A200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SKF 38393 hydrochloride is the hydrochloride salt of SKF-38393, a selective dopamine D1 receptor agonist with a reported IC50 of approximately 110 nM. Supplied as a research-grade solid analytical standard, it is used for in vitro pharmacology, receptor binding, and analytical assays. Not for human or clinical use.

  • Selective D1 receptor agonist, reported IC50 ≈ 110 nM.
  • Hydrochloride salt form improves solubility for biological assays.
  • Molecular weight 291.77 g/mol.
  • Chemical formula C16H18ClNO2.
  • Provided as a solid analytical standard suitable for in vitro studies.
  • Typical purities reported ≥98% by HPLC from third-party suppliers.
  • Not for human or clinical use; for research only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.