Learn More
Cayman Chemical SKF 38393 (hydrobromide), Purity: ≥98%, CAS: 20012-10-6, 50 mg
SDP
Catalog No. 502983014
Encompass_Preferred
A partial agonist of the dopamine D1-like receptors D1 and D5 (Kis = 1 and ~0.5 nM, respectively)
Catalog No. 50-298-3014 Supplier Cayman Chemical Supplier No. 1915050 MG
A partial agonist of the dopamine D1-like receptors D1 and D5 (Kis = 1 and ~0.5 nM, respectively)