Learn More
Enzo Life Sciences SKF-525A. HCl (1g). CAS: 62-68-0
SDP
Supplier:  Enzo Life Sciences BMLEI1051000
Encompass_Preferred
Cytochrome P-450 inhibitor. Inhibits cytochrome P-450-dependent arachidonate metabolism (90% at 50 µM). Calcium antagonist. Blocks glibenclamide-sensitive K+ channels (IC50=4.4mM). Inhibits platelet thromboxane synthesis and stimulates endothelial cell prostacyclin production. Potentiates the effects of many drugs in vivo.  Alternative name: Proadifen. Purity: ≥95% (TLC). Solubility: Soluble in water (25mg/ml) or 100% ethanol. Long Term Storage: Ambient.
Catalog No. 50-201-0548