Learn More
Cayman Chemical SKF 525AhydrochlorIde 100mg
SDP
Catalog No. 502969637
Encompass_Preferred
A widely used, nonspecific CYP450 inhibitor that potentiates the effects of many different different drugs by inhibiting their metabolism (IC50 values in the μM range when tested using human liver microsomes); also acts as a noncompetitive inhibitor of acetylcholine nicotinic receptors
Catalog No. 50-296-9637 Supplier Cayman Chemical Supplier No. 15040100 MG
A widely used, nonspecific CYP450 inhibitor that potentiates the effects of many different different drugs by inhibiting their metabolism (IC50 values in the μM range when tested using human liver microsomes); also acts as a noncompetitive inhibitor of acetylcholine nicotinic receptors