Learn More
Enzo Life Sciences Fluphenazine-N-2-chloroethane . 2HCl (50mg). CAS: 83016-35-7
SDP
Catalog No. 502010388
Encompass_Preferred
A potent irreversible inactivator of calmodulin. Possesses potent antiproliferative and cytotoxic activity. It is also an irreversible D2 dopamine receptor antagonist. Alternative name: SKF-7171A. Purity: ≥98%. Solubility: Soluble in DMSO (50mg/ml) or water (10mg/ml warm). Long Term Storage: Ambient.
Catalog No. 50-201-0388 Supplier Enzo Life Sciences Supplier No. BMLCA3250050
A potent irreversible inactivator of calmodulin. Possesses potent antiproliferative and cytotoxic activity. It is also an irreversible D2 dopamine receptor antagonist. Alternative name: SKF-7171A. Purity: ≥98%. Solubility: Soluble in DMSO (50mg/ml) or water (10mg/ml warm). Long Term Storage: Ambient.