Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SKF-75670 hydrobromide | 62717-63-9 | MFCD11045981 | 99.8% | 350.25 | C17H20BrNO2 | 25MG
SDP

Catalog No. 5000235228
Encompass_Preferred

SKF-75670 hydrobromide is the hydrobromide salt of SKF-75670, a dopamine D1 receptor partial agonist reported to act as a cocaine antagonist. It is supplied for research use in neuroscience and pharmacology, available in solid and ready-to-use solution formats for in vitro assays and receptor profiling.

  • High reported purity (≈99.8%).
  • Available in multiple pack sizes and solution formats for experimental flexibility.
  • Provided as the hydrobromide salt compatible with receptor pharmacology studies.
  • Molecular weight approximately 350.25 g/mol.
  • CAS number 62717-63-9 for unambiguous identification.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-35228 Supplier Medchemexpress LLC Supplier No. HY12504325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SKF-75670 hydrobromide is the hydrobromide salt of SKF-75670, a dopamine D1 receptor partial agonist reported to act as a cocaine antagonist. It is supplied for research use in neuroscience and pharmacology, available in solid and ready-to-use solution formats for in vitro assays and receptor profiling.

  • High reported purity (≈99.8%).
  • Available in multiple pack sizes and solution formats for experimental flexibility.
  • Provided as the hydrobromide salt compatible with receptor pharmacology studies.
  • Molecular weight approximately 350.25 g/mol.
  • CAS number 62717-63-9 for unambiguous identification.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.