Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Skf 81297 hydrobromide | 67287-39-2 | 99.6% | 5 MG
SDP

Catalog No. 5000231470
Encompass_Preferred

SKF 81297 hydrobromide is a selective dopamine D1-like receptor agonist used in pharmacology and neuroscience research for in vitro and in vivo studies.

  • Potent agonist of D1 and D5 dopamine receptors.
  • High purity suitable for research applications.
  • Available in small solid and solution pack sizes for flexible dosing.
  • Off-white to yellow solid appearance.
  • Store solid at 4°C; store solutions at -80°C for long-term storage.
  • Molecular weight 370.67 g/mol; formula C16H17BrClNO2.

Catalog No. 50-002-31470 Supplier Medchemexpress LLC Supplier No. HY122365MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SKF 81297 hydrobromide is a selective dopamine D1-like receptor agonist used in pharmacology and neuroscience research for in vitro and in vivo studies.

  • Potent agonist of D1 and D5 dopamine receptors.
  • High purity suitable for research applications.
  • Available in small solid and solution pack sizes for flexible dosing.
  • Off-white to yellow solid appearance.
  • Store solid at 4°C; store solutions at -80°C for long-term storage.
  • Molecular weight 370.67 g/mol; formula C16H17BrClNO2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.