Learn More
Cayman Chemical SKF 82958 (hydrobromide), Purity: ≥95%, CAS: 74115-01-8, 5 mg
SDP
Catalog No. 502940108
Encompass_Preferred
A dopamine D1 receptor agonist (K0.5 = 4 nM in a radioligand binding assay); selective for dopamine D1 over D2 receptors (K0.5 = 73 nM); induces dopamine D1 receptor-dependent adenylate cyclase activity in rat striatal membranes (EC50 = 491 nM); prevents and reverses fentanyl-induced respiratory depression in cats at 3 mg/kg; increases locomotion and rearing in rats
Catalog No. 50-294-0108 Supplier Cayman Chemical Supplier No. 290335 MG
A dopamine D1 receptor agonist (K0.5 = 4 nM in a radioligand binding assay); selective for dopamine D1 over D2 receptors (K0.5 = 73 nM); induces dopamine D1 receptor-dependent adenylate cyclase activity in rat striatal membranes (EC50 = 491 nM); prevents and reverses fentanyl-induced respiratory depression in cats at 3 mg/kg; increases locomotion and rearing in rats