Learn More
Medchemexpress LLC SKF-83566 hydrobromide | 99295-33-7 | MFCD04975449 | ≥98.0% | 332.23 g/mol | C17H18BrNO | 25 MG
SDP
Catalog No. 5000244863
Encompass_Preferred
SKF-83566 hydrobromide is the hydrobromide salt of a D1/D5 dopamine receptor antagonist used in neuroscience research. It is blood-brain permeable, orally active, and is commonly applied in studies of dopaminergic signaling, Parkinson's disease models, and nicotine craving.
  • Potent D1/D5 receptor antagonist with activity at related targets.
  • Also shows weaker antagonism at 5-HT2 receptors and inhibitory activity at the dopamine transporter at higher concentrations.
  • Blood-brain permeable and orally active, suitable for in vivo studies.
  • Typical purity ≥98.0% (HPLC).
  • Molecular formula C17H18BrNO and molecular weight 332.23 g/mol.
  • Supplied as a 25 mg package for research use.
Catalog No. 50-002-44863 Supplier Medchemexpress LLC Supplier No. HY103430A25MG
SKF-83566 hydrobromide is the hydrobromide salt of a D1/D5 dopamine receptor antagonist used in neuroscience research. It is blood-brain permeable, orally active, and is commonly applied in studies of dopaminergic signaling, Parkinson's disease models, and nicotine craving.
  • Potent D1/D5 receptor antagonist with activity at related targets.
  • Also shows weaker antagonism at 5-HT2 receptors and inhibitory activity at the dopamine transporter at higher concentrations.
  • Blood-brain permeable and orally active, suitable for in vivo studies.
  • Typical purity ≥98.0% (HPLC).
  • Molecular formula C17H18BrNO and molecular weight 332.23 g/mol.
  • Supplied as a 25 mg package for research use.