Learn More
Abcam SKF 83566 hydrobromide, D1 receptor antagonist, 10MG
Catalog No. 502706850
Encompass_Preferred
MW 413.1 Da, Purity >99%. Potent, selective D₁ receptor antagonist (Ki = 0.4 nM). Centrally active following systemic administration. Various neurological effects in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6850 Supplier Abcam Supplier No. AB14114610MG
MW 413.1 Da, Purity >99%. Potent, selective D₁ receptor antagonist (Ki = 0.4 nM). Centrally active following systemic administration. Various neurological effects in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement