Learn More
Cayman Chemical SKF 83566hydrobromIde 10mg
SDP
Catalog No. 502990039
Encompass_Preferred
A dopamine D1 receptor antagonist; reduces dexbenzetimide- and scopolamine-induced locomotor stereotypy in rats at 0.005, 0.01, and 0.03 mg/kg; decreases ghrelin-induced increases in novel object exploration and food intake in rats at 100 UG/kg; reduces schedule-induced polydipsia in rats
Catalog No. 50-299-0039 Supplier Cayman Chemical Supplier No. 2948010 MG
A dopamine D1 receptor antagonist; reduces dexbenzetimide- and scopolamine-induced locomotor stereotypy in rats at 0.005, 0.01, and 0.03 mg/kg; decreases ghrelin-induced increases in novel object exploration and food intake in rats at 100 UG/kg; reduces schedule-induced polydipsia in rats