Learn More
Medchemexpress LLC Skf-83959 hydrobromide | 67287-95-0 | 99.9% | 398.72 | C18H21BrClNO2 | 5 MG
SDP
Catalog No. 5000245212
Encompass_Preferred
SKF-83959 hydrobromide is the hydrobromide salt of SKF-83959, a dopamine D1-like receptor partial agonist used in neuropharmacology research. Supplied as a white to off-white solid with high purity for in vitro and in vivo studies. Recommended storage is sealed at -20°C and protected from moisture; in solution, short-term storage at -20°C or -80°C is indicated.
  • High purity suitable for research applications
  • Solid form simplifies handling and storage
  • Active at dopamine D1-like receptors for pharmacological studies
  • Stable when stored sealed at low temperature
  • Available in small 5 mg quantity for screening and early-stage experiments
Catalog No. 50-002-45212 Supplier Medchemexpress LLC Supplier No. HY1034125MG
SKF-83959 hydrobromide is the hydrobromide salt of SKF-83959, a dopamine D1-like receptor partial agonist used in neuropharmacology research. Supplied as a white to off-white solid with high purity for in vitro and in vivo studies. Recommended storage is sealed at -20°C and protected from moisture; in solution, short-term storage at -20°C or -80°C is indicated.
  • High purity suitable for research applications
  • Solid form simplifies handling and storage
  • Active at dopamine D1-like receptors for pharmacological studies
  • Stable when stored sealed at low temperature
  • Available in small 5 mg quantity for screening and early-stage experiments