Learn More
SKF 83959 hydrobromide, Tocris Bioscience™
Description
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 398.73 |
| Percent Purity | >98% |
| Chemical Name or Material | SKF 83959 hydrobromide |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.