Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKF 83959 hydrobromide, Tocris Bioscience™

Catalog No. 207450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-745-0 50 mg
20-741-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-745-0 Supplier R&D Systems Supplier No. 2074/50
This item is not returnable. View return policy

D1-like partial agonist

Dopamine D1-like receptor partial agonist (Ki values are 1.18, 7.56, 920 and 399 nM for rat D1, D5, D2 and D3 receptors respectively). May act as an antagonist in vivo, producing anti-Parkinsonian effects and antagonizing the behavioral effects of cocaine. Shown to potentiate agonist binding of the σ1 receptor.

Chemical Identifiers

CAS 67287-95-0
Molecular Formula C18H21BrClNO2
Molecular Weight (g/mol) 398.725
InChI Key FHYWNBUFNGHNCP-UHFFFAOYSA-N
Synonym skf 83959 hydrobromide, skf-83959 hydrobromide, 6-chloro-7,8-dihydroxy-3-methyl-1-3-methylphenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrobromide, 6-chloro-3-methyl-1-3-methylphenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, 6-chloro-2,3,4,5-tetrahydro-3-methyl-1-3-methylphenyl-1h-3-benzazepine-7,8-diol hydrobromide, 1h-3-benzazepine-7,8-diol, 6-chloro-2,3,4,5-tetrahydro-3-methyl-1-3-methylphenyl-, hydrobromide, 1h-3-benzazepine-7,8-diol, 6-chloro-2,3,4,5-tetrahydro-3-methyl-1-3-methylphenyl-, hydrobromide 1:1, skf-83959 hydrobromide hplc , solid, 6-chloro-2,3,4,5-tetrahydro-3-methyl-1-3-methylphenyl-1h-3-benzazepine-7,8-diol, 6-chloro-7,8-dihydroxy-3-methyl-1-3-methylphenyl-2,3,4,5-tetrahydro-1h-3-benzazepinium bromide
PubChem CID 11957685
ChEBI CHEBI:63996
IUPAC Name 9-chloro-3-methyl-5-(3-methylphenyl)-1,2,4,5-tetrahydro-3-benzazepine-7,8-diol;hydrobromide
SMILES CC1=CC=CC(=C1)C2CN(CCC3=C(C(=C(C=C23)O)O)Cl)C.Br

Specifications

Quantity 50 mg
Formula Weight 398.73
Percent Purity >98%
Chemical Name or Material SKF 83959 hydrobromide
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.