Learn More
Cayman Chemical SKF 96365 (hydrochloride), Purity: ≥98%, CAS: 130495-35-1, 1 mg
SDP
Catalog No. 502890939
Encompass_Preferred
Inhibits the receptor-mediated influx of calcium via voltage-gated calcium channels (IC50 = 10 µM); inhibits the acetylcholine-induced depolarization of circular smooth muscle in a dose-dependent manner at 3-50 µM
Catalog No. 50-289-0939 Supplier Cayman Chemical Supplier No. 100093121 MG
Inhibits the receptor-mediated influx of calcium via voltage-gated calcium channels (IC50 = 10 µM); inhibits the acetylcholine-induced depolarization of circular smooth muscle in a dose-dependent manner at 3-50 µM