Learn More
Abcam SKF 96365, TRP channel blocker, 10MG
Catalog No. 502707721
Encompass_Preferred
MW 402.9 Da, Purity >99%. Blocker of TRP cation channels. Inhibits capacitative Ca²+ entry.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7721 Supplier Abcam Supplier No. AB12028010MG
MW 402.9 Da, Purity >99%. Blocker of TRP cation channels. Inhibits capacitative Ca²+ entry.
The product is subject to the following: Abcam Restricted Use Statement