Learn More
Abcam SKF 97541, Highly GABAB agonist, 10MG
Supplier:  Abcam AB12025010MG
Encompass_Preferred
MW 137.12 Da, Purity >98%. Highly potent GABAB agonist, 10-fold more potent than Baclofen (ab120149). (EC50 values for depression of striatal synaptic potentials are 920 nM and 1.25 μM for SKF97541 and baclofen, respectively). Also available in simple stock solutions (ab146688) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7744