Learn More
MSC SKF Ball Transfer Unit, 25.4 mm Ball, Carbon Steel, Oval Base
Catalog No. 5000516921
Encompass
This ball transfer unit provides multi-directional material handling for moving heavy loads smoothly and precisely with minimal effort. Featuring a 25.4 mm carbon steel ball within an oval base, it is engineered for high load capacity applications, supporting up to 121 lb. The unit's design includes a large load-bearing ball supported by smaller balls within a hemispherical cup, and its housing can incorporate a seal for cleaning the main ball during rotation.
  • High load capacity
  • Multi-directional movement capability
  • Smooth and precise material handling
  • Reduced friction
  • Standard dirt exit hole
  • Drill hole fixing for secure installation
  • Can be used in any orientation
Catalog No. 50-005-16921 Supplier MSC Supplier No. 06377485
This ball transfer unit provides multi-directional material handling for moving heavy loads smoothly and precisely with minimal effort. Featuring a 25.4 mm carbon steel ball within an oval base, it is engineered for high load capacity applications, supporting up to 121 lb. The unit's design includes a large load-bearing ball supported by smaller balls within a hemispherical cup, and its housing can incorporate a seal for cleaning the main ball during rotation.
  • High load capacity
  • Multi-directional movement capability
  • Smooth and precise material handling
  • Reduced friction
  • Standard dirt exit hole
  • Drill hole fixing for secure installation
  • Can be used in any orientation