Learn More
Medchemexpress LLC HY-100228A 5mg Medchemexpress, SKF89976A (hydrochloride) CAS:85375-15-1 Purity:>98%
SDP
Catalog No. 501931764
Encompass_Preferred
Medchemexpress, HY-100228A 5mg SKF89976A (hydrochloride) CAS:85375-15-1 SKF89976A hydrochloride is a selective GABA transporter ( GAT-1 ) inhibitor with IC 50 s of 0.28 μM, 137.34 μM and 202.8 μM for GAT-1, GAT-2 and GAT-3 in CHO cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1764 Supplier Medchemexpress LLC Supplier No. HY100228A5MG
Add to Cart
Add to Cart
Medchemexpress, HY-100228A 5mg SKF89976A (hydrochloride) CAS:85375-15-1 SKF89976A hydrochloride is a selective GABA transporter ( GAT-1 ) inhibitor with IC 50 s of 0.28 μM, 137.34 μM and 202.8 μM for GAT-1, GAT-2 and GAT-3 in CHO cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.