Learn More
Medchemexpress LLC SKF89976A hydrochloride | 85375-15-1 | MFCD01321071 | 98.7% | 371.90 g/mol | C22H26ClNO2 | 1 ML
SDP
Catalog No. 5000215749
Encompass_Preferred
SKF89976A hydrochloride is the hydrochloride salt of SKF89976A, a selective GABA transporter 1 (GAT-1) inhibitor used in neuroscience and pharmacology research to probe GABA uptake and transporter function. The compound is supplied in solid form and as ready-to-use 10 mM solutions in DMSO, and is accompanied by SDS, data sheet, and certificate of analysis documentation.
  • Selective inhibitor of GABA transporter 1 (GAT-1) used to study GABA uptake.
  • Documented IC50 values for GAT-1, GAT-2, and GAT-3 in CHO cells.
  • Available as solid and 10 mM solution in DMSO for ready use.
  • High reported purity suitable for biochemical assays.
  • Accompanied by SDS, data sheet, and COA for handling and quality assurance.
Catalog No. 50-002-15749 Supplier Medchemexpress LLC Supplier No. HY100228A10MM 1ML
SKF89976A hydrochloride is the hydrochloride salt of SKF89976A, a selective GABA transporter 1 (GAT-1) inhibitor used in neuroscience and pharmacology research to probe GABA uptake and transporter function. The compound is supplied in solid form and as ready-to-use 10 mM solutions in DMSO, and is accompanied by SDS, data sheet, and certificate of analysis documentation.
  • Selective inhibitor of GABA transporter 1 (GAT-1) used to study GABA uptake.
  • Documented IC50 values for GAT-1, GAT-2, and GAT-3 in CHO cells.
  • Available as solid and 10 mM solution in DMSO for ready use.
  • High reported purity suitable for biochemical assays.
  • Accompanied by SDS, data sheet, and COA for handling and quality assurance.