Learn More
Lonza Walkersville SKGM - 2 Skeletal Muscle Cell Growth Medium-2 SingleQuots Supplements and Growth Factors
Catalog No. NC9993178
Encompass
SkGM -2 SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the SkGM -2 Skeletal Muscle Myoblast Medium BulletKit (Catalog Number CC-3245). The full SkGM -2 BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3245. We offers two varieties of Skeletal Muscle Growth Medium - SkGM and SkGM-2.SkGM-2 Skeletal Muscle Cell Growth Medium-2 is an enhancement of SkGM Medium. Fetuin has been removed and there has been further optimization of the basal media.SkGM Medium is optimized for use with human skeletal muscle cells. SkGM -2 is specially designed for use with normal and diabetic skeletal muscle myoblasts.Content:1 x Red Cap Vial with GA-1000 0.50 mL1 x Green Cap Vial with hEGF 0.50 mL1 x Natural Cap Vial with Dexamethasone 0.50 mL1 x Bottle with FBS 50.00 mL1 x Bottle L-Glutamine 10.00 mL
Catalog No. NC9993178 Supplier Lonza Walkersville Supplier No. CC3244
SkGM -2 SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the SkGM -2 Skeletal Muscle Myoblast Medium BulletKit (Catalog Number CC-3245). The full SkGM -2 BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3245. We offers two varieties of Skeletal Muscle Growth Medium - SkGM and SkGM-2.SkGM-2 Skeletal Muscle Cell Growth Medium-2 is an enhancement of SkGM Medium. Fetuin has been removed and there has been further optimization of the basal media.SkGM Medium is optimized for use with human skeletal muscle cells. SkGM -2 is specially designed for use with normal and diabetic skeletal muscle myoblasts.Content:1 x Red Cap Vial with GA-1000 0.50 mL1 x Green Cap Vial with hEGF 0.50 mL1 x Natural Cap Vial with Dexamethasone 0.50 mL1 x Bottle with FBS 50.00 mL1 x Bottle L-Glutamine 10.00 mL