Learn More
ATCC SK-HEP-1; Liver Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502382523
Encompass
SK-HEP-1 are endothelial cells that were isolated from the liver of a 52-year-old, White male patient with adenocarcinoma. This product has applications in cardiovascular disease research, toxicology, and immuno-oncology.
Catalog No. 50-238-2523 Supplier ATCC Supplier No. HTB52
SK-HEP-1 are endothelial cells that were isolated from the liver of a 52-year-old, White male patient with adenocarcinoma. This product has applications in cardiovascular disease research, toxicology, and immuno-oncology.