Learn More
Medchemexpress LLC SKI-178 | 1259484-97-3 | 99.3% | 50 MG
SDP
Catalog No. 5000339143
Encompass_Preferred
SKI-178 is a potent inhibitor of both sphingosine kinase-1 (SphK1) and sphingosine kinase-2 (SphK2). It demonstrates cytotoxicity in various cancer cell lines, including agent-sensitive and multi-agent resistant types, with IC50 concentrations ranging from 0.1 to 1.8 μM. This compound has been shown to induce apoptosis in human acute myeloid leukemia cell lines in a CDK1-dependent manner and correlates with prolonged Bcl-2 phosphorylation. In in-vivo studies, SKI-178 inhibited leukemic progression in the MLL-AF9 mouse model, leading to a decrease in white blood cell counts.
  • Potent SphK1 and SphK2 inhibitor
  • Exhibits cytotoxicity in various cancer cell lines
  • Induces apoptosis in human acute myeloid leukemia cells
  • Correlates with prolonged Bcl-2 phosphorylation
  • Inhibits leukemic progression in MLL-AF9 mouse model
Catalog No. 50-003-39143 Supplier Medchemexpress LLC Supplier No. HY1289250MG
SKI-178 is a potent inhibitor of both sphingosine kinase-1 (SphK1) and sphingosine kinase-2 (SphK2). It demonstrates cytotoxicity in various cancer cell lines, including agent-sensitive and multi-agent resistant types, with IC50 concentrations ranging from 0.1 to 1.8 μM. This compound has been shown to induce apoptosis in human acute myeloid leukemia cell lines in a CDK1-dependent manner and correlates with prolonged Bcl-2 phosphorylation. In in-vivo studies, SKI-178 inhibited leukemic progression in the MLL-AF9 mouse model, leading to a decrease in white blood cell counts.
  • Potent SphK1 and SphK2 inhibitor
  • Exhibits cytotoxicity in various cancer cell lines
  • Induces apoptosis in human acute myeloid leukemia cells
  • Correlates with prolonged Bcl-2 phosphorylation
  • Inhibits leukemic progression in MLL-AF9 mouse model