Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SKI-I 100mg | 306301-68-8 | 406.44 | C25H18N4O2 | 100 MG
SDP

Catalog No. 5000227568
Encompass_Preferred

SKI-I is a small-molecule sphingosine kinase inhibitor used in biochemical and cellular research to probe sphingolipid signaling and apoptosis pathways. It is supplied as a purified research reagent with stated solubility and storage recommendations for laboratory use only.

  • Research-grade sphingosine kinase inhibitor for biochemical and cellular studies.
  • High reported purity of 99.14%.
  • Molecular weight 406.44 and formula C25H18N4O2.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Available in multiple pack sizes, including 100 MG.

Catalog No. 50-002-27568 Supplier Medchemexpress LLC Supplier No. HY115735100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SKI-I is a small-molecule sphingosine kinase inhibitor used in biochemical and cellular research to probe sphingolipid signaling and apoptosis pathways. It is supplied as a purified research reagent with stated solubility and storage recommendations for laboratory use only.

  • Research-grade sphingosine kinase inhibitor for biochemical and cellular studies.
  • High reported purity of 99.14%.
  • Molecular weight 406.44 and formula C25H18N4O2.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Available in multiple pack sizes, including 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.