Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SKI-I 10mg | 306301-68-8 | 406.44 | C25H18N4O2 | 10 MG
SDP

Catalog No. 5000307758
Encompass_Preferred

SKI-I is a small-molecule sphingosine kinase inhibitor supplied as a 10 mg research-grade solid for laboratory use. It inhibits human sphingosine kinase with reported IC50 ≈ 1.2 μM and shows additional activity against hERK2; reported purity and storage conditions support reproducible experimental use.

  • High reported purity (99.14%).
  • Off-white to light yellow solid; convenient 10 mg quantity for assay development.
  • Soluble in DMSO at 100 mg/mL; suitable for in vitro use and formulatable for in vivo studies.
  • Storage: powder at 4°C protected from light; aliquots in solvent stable at -80°C for up to 6 months.
  • Demonstrated cytotoxicity against multiple human cancer cell lines at low micromolar concentrations.

Catalog No. 50-003-07758 Supplier Medchemexpress LLC Supplier No. HY11573510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SKI-I is a small-molecule sphingosine kinase inhibitor supplied as a 10 mg research-grade solid for laboratory use. It inhibits human sphingosine kinase with reported IC50 ≈ 1.2 μM and shows additional activity against hERK2; reported purity and storage conditions support reproducible experimental use.

  • High reported purity (99.14%).
  • Off-white to light yellow solid; convenient 10 mg quantity for assay development.
  • Soluble in DMSO at 100 mg/mL; suitable for in vitro use and formulatable for in vivo studies.
  • Storage: powder at 4°C protected from light; aliquots in solvent stable at -80°C for up to 6 months.
  • Demonstrated cytotoxicity against multiple human cancer cell lines at low micromolar concentrations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.